S. pneumoniae Recombinant Streptococcus pneumoniae type 12f Capsular Polysaccharide Antigen
Catalog # 77682-720
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:S. pneumoniae
- Protein Synonyms:Streptococcus pneumoniae type 12f Capsular Polysaccharide Antigen
- Protein/Peptide Name:Streptococcus pneumoniae type 12f Capsular Polysaccharide Antigen
- Endotoxin Level:Very low
- Shipping Temperature:Dry Ice
- Cat. No.:77682-720
Specifications
About this item
Recombinant Streptococcus pneumoniae type 12f Capsular Polysaccharide antigen.