HSV-2 Recombinant gB (from E. coli)
Catalog # 77682-706
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:HSV-2
- Protein Synonyms:gB|gB immunodominant regions
- Protein/Peptide Name:gB
- Purity:Protein is >90% pure as determined by SDS PAGE.
- Molecular Weight:82 kDa
- Endotoxin Level:Very low
- Formulation:10 mM Phosphate buffer pH 7.6 and 75 mM NaCl.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-706
Specifications
About this item
Recombinant HSV-2 gB antigen, his-tagged.