RSV Recombinant Respiratory Syndrome Virus (from E. coli)
Catalog # 77682-698
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:RSV
- Protein Synonyms:Respiratory Syndrome Virus|RSV
- Protein/Peptide Name:Respiratory Syndrome Virus
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:It has a predicted MW of 38.6 kDa.
- Endotoxin Level:Very low
- Formulation:Supplied from sterile 10 mM PBS Buffer, pH 7.4.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-698
Specifications
About this item
Recombinant Respiratory Syndrome Virus antigen.