Hepatitis B virus (HBV) Recombinant hepatitis B virus e antigen (from E. coli)
Catalog # 77682-682
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Hepatitis B virus (HBV)
- Protein Synonyms:HBV|HBeAg|hepatitis B virus e antigen|hepatitis B virus
- Protein/Peptide Name:hepatitis B virus e antigen
- Purity:Protein is >95% pure as determined by 12% PAGE (coomassie staining).
- Molecular Weight:18 kDa
- Endotoxin Level:Very low
- Formulation:Phosphate buffered saline with 50 mM arginine.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-682
Specifications
About this item
Recombinant hepatitis B virus e antigen, his-tagged.