HIV2 Recombinant gp36 (from E. coli)
Catalog # 77682-678
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:HIV2
- Protein Synonyms:gp36|gp36 immunodominant regions
- Protein/Peptide Name:gp36
- Purity:Greater than 95.0% as determined by HPLC analysis and SDS-PAGE.
- Molecular Weight:34 kDa
- Endotoxin Level:Very low
- Formulation:0.01M Na2CO3, 0.01M Na3EDTA, 0.014 Mb-mercaptoethanol, 0.05% Tween-20.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-678
Specifications
About this item
Recombinant HIV-2 gp36 antigen, beta-galactosidase-tagged.