HIV2 Recombinant gp36 (from Synthetic)
Catalog # 77682-676
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:Synthetic
- Species:HIV2
- Protein Synonyms:gp36|gp36 immunodominant regions
- Protein/Peptide Name:gp36
- Purity:Greater than 95.0% as determined by HPLC.
- Endotoxin Level:Very low
- Formulation:(1 mg/1 ml) in H2O.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-676
Specifications
About this item
Recombinant HIV-2 gp36 antigen.