Norovirus Recombinant Capsid P-Domain (from E. coli)
Catalog # 77682-652
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Norovirus
- Protein Synonyms:Capsid P-Domain|Capsid
- Protein/Peptide Name:Capsid P-Domain
- Purity:Protein is >95% pure as determined by 12% PAGE (coomassie staining).
- Molecular Weight:30 kDa
- Endotoxin Level:Very low
- Formulation:Phosphate buffer and 17 mM K2CO3.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-652
Specifications
About this item
Recombinant Norovirus Group-2 Capsid P-Domain antigen, his-tagged.