HIV-1 Recombinant p66 (from Baculovirus (Insect cells))
Catalog # 77682-650
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:HIV-1
- Protein Synonyms:p66 pol|p66|p66 immunodominant regions
- Protein/Peptide Name:p66
- Purity:Greater than 90.0% as determined by HPLC analysis & SDS-PAGE.
- Molecular Weight:71 kDa
- Endotoxin Level:Very low
- Formulation:The protein solution contains 30 mM Tris pH-7, 0.15 M NaCl and 0.2 mM EDTA.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-650
Specifications
About this item
Recombinant HIV-1 p66 pol antigen.