HIV-1 Recombinant p55 (from Baculovirus (Insect cells))
77682-648
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:HIV-1
- Protein Synonyms:p55|p55 gag|p55 immunodominant regions
- Protein/Peptide Name:p55
- Purity:Greater than 90.0% as determined by HPLC analysis & SDS-PAGE.
- Molecular Weight:55 kDa
- Endotoxin Level:Very low
- Formulation:The HIV-1 p55 gag protein solution contains 10 mM Tris pH 8, 140 mM NaCl and 400 mM L-Arginine.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-648
Recombinant HIV-1 p55 gag antigen.