HIV-1 Recombinant p24 (from E. coli)
Catalog # 77682-646
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:HIV-1
- Protein Synonyms:p24|p24 immunodominant regions
- Protein/Peptide Name:p24
- Purity:Greater than 90.0% as determined by HPLC analysis and SDS-PAGE.
- Endotoxin Level:Very low
- Formulation:1.5 M urea, 25 mM Tris-HCl pH-8.0 and 50% Glycerol.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-646
Specifications
About this item
Recombinant HIV-1 p24 antigen, GST-tagged, HRP-Labled.