Order Entry
United States
Orders LinkContactUsLinkComponent
HIV-1 Recombinant p24 (from E. coli)

HIV-1 Recombinant p24 (from E. coli)

Vial
Catalog # 77682-644
Supplier:   Creative Biomart
Supplier #:  HIV-281
HIV-1 Recombinant p24 (from E. coli)
Catalog # 77682-644
Supplier:  Creative Biomart
CAS Number:  
HIV-1 Recombinant p24 (from E. coli)
HIV-1 Recombinant p24 (from E. coli)

Vial

CAS Number:  
Price
$1,022.60
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Recombinant HIV-1 p24 antigen, his-tagged.

Specifications

Specifications

  • Pack type:
    Vial
  • Cat. No.:
    77682-644
  • Molecular Weight:
    21.2 kDa
  • Shipping Temperature:
    Dry Ice
  • Protein/Peptide Type:
    Recombinant
  • Formulation:
    The HIV-1 p24 protein solution contains 20 mM Tris-HCl pH-8, 0.1 mM PMSF, 0.1 M NaCl and 10% glycerol.
  • Endotoxin Level:
    Very low
  • Purity:
    Greater than 80.0% as determined by SDS-PAGE.
  • Protein Synonyms:
    p24|p24 immunodominant regions
  • Protein/Peptide Name:
    p24
  • Species:
    HIV-1
  • Source:
    E. coli

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.