Order Entry
United States
ContactUsLinkComponent
SARS Recombinant nucleocapsid phosphoprotein
SARS Recombinant nucleocapsid phosphoprotein
Catalog # 77682-630
Supplier:  Creative Biomart
New Product
CAS Number:  
undefined
SARS Recombinant nucleocapsid phosphoprotein
Catalog # 77682-630
Supplier:  Creative Biomart
Supplier Number:  HIV-274
New Product
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Protein/Peptide Type:
    Recombinant
  • Species:
    SARS
  • Protein Synonyms:
    structural protein|nucleocapsid phosphoprotein|N|ORF9|SARS Core
  • Protein/Peptide Name:
    nucleocapsid phosphoprotein
  • Purity:
    SARS Core protein is >95% pure as determined by 10% PAGE (coomassie staining).
  • Molecular Weight:
    35 kDa
  • Endotoxin Level:
    Very low
  • Formulation:
    50 mM Tris-HCl, pH-8, 60 mM NaCl and 50% glycerol.
  • Shipping Temperature:
    Dry Ice
  • Cat. No.:
    77682-630

Specifications

About this item

Recombinant SARS Core antigen.