Hepatitis B virus (HBV) Recombinant Pre-S (from E. coli)
77682-324
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Hepatitis B virus (HBV)
- Protein Synonyms:Pre-S|adw2|HBV Pre-S
- Protein/Peptide Name:Pre-S
- Purity:Protein is >95% pure as determined by 12% PAGE (coomassie staining)
- Molecular Weight:25 kDa
- Endotoxin Level:very low
- Formulation:Sterile Filtered solution containing PBS, 1 M urea and 0.05% sodium azide.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-324
Recombinant HBV Pre-S Antigen, His-tagged.