CoV-2 Recombinant Membrane (from E. coli)
77682-312
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:CoV-2
- Protein Synonyms:E|SARS Membrane|Envelope|M|Membrane
- Protein/Peptide Name:Membrane
- Purity:Protein is >90% pure as determined SDS-page
- Molecular Weight:34.2 kDa
- Endotoxin Level:very low
- Formulation:CoV-2 Membrane Envelope protein solution is supplied in 1XPBS.
- Shipping Temperature:Dry Ice
- Cat. No.:77682-312
Recombinant CoV-2 Membrane Env Antigen, His-tagged.