Trypanosoma cruzi Recombinant Chagas 1F8 (from E. coli)
Catalog # 77682-298
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Trypanosoma cruzi
- Protein Synonyms:Chagas 1F8|Epitope of the flagellar calcium-binding protein FCABP|FcaBP
- Protein/Peptide Name:Chagas 1F8
- Purity:Protein is >90% pure as determined by SDS page
- Molecular Weight:25.8 kDa
- Endotoxin Level:very low
- Formulation:1F8 Chagas antigen was lyopH ilized from 20 mM Hepes, 300 mM NaCl, pH 8, and varying 284 mM imidazole.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-298
Specifications
About this item
Recombinant Trypanosoma cruzi 1F8 Chagas Antigen, His-tagged.