Order Entry
United States
Orders LinkContactUsLinkComponent
EBV Recombinant p23

EBV Recombinant p23

Vial
Catalog # 77682-288
Supplier:   Creative Biomart
Supplier #:  EBV-059
EBV Recombinant p23
Catalog # 77682-288
Supplier:  Creative Biomart
CAS Number:  
EBV Recombinant p23
EBV Recombinant p23

Vial

CAS Number:  
Price
$1,022.60
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Recombinant EBV p23 Antigen, His-tagged.

Specifications

Specifications

  • Pack type:
    Vial
  • Cat. No.:
    77682-288
  • Molecular Weight:
    17.7 kDa
  • Shipping Temperature:
    Dry Ice
  • Protein/Peptide Type:
    Recombinant
  • Formulation:
    1X PBS pH 7.5 and 300 mM Imidazole.
  • Endotoxin Level:
    very low
  • Purity:
    Protein is >95% pure as determined by 10% PAGE (coomassie staining)
  • Protein Synonyms:
    p23|p23 immunodominant regions
  • Protein/Peptide Name:
    p23
  • Grade:
    Ultrapure grade
  • Species:
    EBV (Epstein-Barr virus)

Specifications