Influenza B Recombinant Hemagglutinin
Catalog # 77682-264
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:Influenza B
- Protein Synonyms:Influenza B virus|Hemagglutinin|Influenza B virus B/Phuket/3073/2013 Hemagglutinin (formaldehyde inactivated)|Influenza virus type B
- Protein/Peptide Name:Hemagglutinin
- Endotoxin Level:very low
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-264
Specifications
About this item
Recombinant Influenza B virus B/Phuket/3073/2013 Hemagglutinin Antigen (formaldehyde inactivated).