Dengue Virus Recombinant non-structural protein 1
77682-262
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:DENV
- Protein Synonyms:Non structural protein 1|non-structural protein 1|NS1|nonstructural antigen 1
- Protein/Peptide Name:non-structural protein 1
- Purity:Protein is >95% pure as determined by 12% PAGE (coomassie staining)
- Molecular Weight:45 kDa
- Endotoxin Level:very low
- Formulation:Phosphate buffered saline and 50 mM arginine.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-262
Recombinant Dengue Subtype 4 NS1 Antigen, His-tagged.