Dengue Virus Recombinant non-structural protein 1
Catalog # 77682-244
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:DENV
- Protein Synonyms:Non structural protein 1|non-structural protein 1|NS1|nonstructural antigen 1
- Protein/Peptide Name:non-structural protein 1
- Purity:Protein is >90% pure as determined by 10% SDS-PAGE (coomassie staining)
- Molecular Weight:45 kDa
- Endotoxin Level:very low
- Formulation:PBS, 25 mM arginine and 1M urea.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-244
Specifications
About this item
Recombinant Dengue NS1 Antigen, His-tagged.