Dengue Virus Recombinant envelope protein (from Insect Cells)
77682-236
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:DENV
- Protein Synonyms:E|Envelope
- Protein/Peptide Name:envelope protein
- Purity:Protein is >95% pure as determined by 12.5% SDS-page
- Molecular Weight:50 kDa
- Endotoxin Level:very low
- Formulation:Dengue Envelope-4 protein solution in 1X D-PBS, pH 7.4, 0.1% Thimerosal, 5 mM EDTA, 1 µg/ml of Leupeptin, Aprotinin and Pepstatin A.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-236
Recombinant Dengue Type 4 Envelope Antigen.