Dengue Virus Recombinant envelope protein (from E. coli)
77682-204
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:DENV
- Protein Synonyms:E|Envelope
- Protein/Peptide Name:envelope protein
- Purity:Protein is >95% pure as determined by 12% PAGE (coomassie staining)
- Molecular Weight:32 kDa
- Endotoxin Level:very low
- Formulation:pH ospH ate buffered saline, pH 7.4.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-204
Recombinant Dengue Type 1 Envelope Antigen, His-tagged.