CoV-2 Recombinant surface glycoprotein (from HEK293 Cells)
Catalog # 77682-172
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:CoV-2
- Protein Synonyms:structural protein|Spike glycoprotein|surface glycoprotein|spike protein|CoV-2 S1|E2|S|spike glycoprotein|Human coronavirus spike glycoprotein|Spike S1|S glycoprotein|E2 glycoprotein
- Protein/Peptide Name:surface glycoprotein
- Purity:Protein is >95% pure as determined SDS-page
- Endotoxin Level:very low
- Formulation:CoV-S1 RBD proteinis lyophilized from 1x PBS pH 7.4 + 5% trehalose.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-172
Specifications
About this item
Recombinant CoV-2-S1 Protein, Fc-tagged.