CoV-2 Recombinant surface glycoprotein (from E. coli)
Catalog # 77682-162
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:CoV-2
- Protein Synonyms:structural protein|Spike glycoprotein|surface glycoprotein|spike protein|E2|S|spike glycoprotein|Human coronavirus spike glycoprotein|S glycoprotein|E2 glycoprotein
- Protein/Peptide Name:surface glycoprotein
- Purity:CoV 2019 Spike S1 Proteinis >90% pure as determined SDS-page
- Endotoxin Level:very low
- Formulation:CoV 2019 Spike S1 (200 - 800 a a)Protein 1 mg/ml solution is supplied in1X PBS.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-162
Specifications
About this item
Recombinant CoV-2 Spike S1 (200-800) Protein, His-tagged.