CoV-2 Recombinant surface glycoprotein (from HEK293 Cells)
Catalog # 77682-146
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:CoV-2
- Protein Synonyms:structural protein|Spike glycoprotein|surface glycoprotein|spike protein|CoV-2 S1|E2|S|spike glycoprotein|Human coronavirus spike glycoprotein|Spike S1|S glycoprotein|E2 glycoprotein
- Protein/Peptide Name:surface glycoprotein
- Purity:Protein is >90% pure as determined SDS-page
- Molecular Weight:26.5 kDa
- Endotoxin Level:very low
- Formulation:Lyophilized from a 0.2 μm filtered concentrated solution in PBS pH 7.4 and 10% treaholse.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-146
Specifications
About this item
Recombinant CoV-2 S1 (319-537) Protein, His-tagged.