CoV-2 Recombinant nucleocapsid phosphoprotein (from E. coli)
77682-138
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:CoV-2
- Protein Synonyms:Nucleoprotein|structural protein|nucleocapsid phosphoprotein|2019-nCoV nucleocapsid protein|N|NC|ORF9|Protein N
- Protein/Peptide Name:nucleocapsid phosphoprotein
- Purity:CoV 2019 Nucleocapsid protein is >95% pure as determined SDS-page
- Molecular Weight:48 kDa
- Endotoxin Level:very low
- Formulation:CoV 2019 protein solution is supplied in PBS and 25 mM K₂CO₃.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-138
Recombinant CoV-2 Nucleocapsid Protein, His-tagged.