Cytomegalovirus (CMV) Recombinant Pp65 (UL83) immunodominant regions (from E. coli)
77682-120
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Cytomegalovirus (CMV)
- Protein Synonyms:Pp65 immunodominant regions|Pp65|UL83|Pp65 (UL83) immunodominant regions
- Protein/Peptide Name:Pp65 (UL83) immunodominant regions
- Purity:CMV Pp65 protein is >90% pure as determined by SDS-page
- Molecular Weight:52.2 kDa
- Endotoxin Level:very low
- Formulation:CMV PP65 protein solution is supplied in PBS.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-120
Recombinant CMV Pp65 Antigen, GST-tagged.