Influenza A Recombinant Hemagglutinin
Catalog # 77682-110
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:Influenza A
- Protein Synonyms:Influenza A virus A/California/7/2009 (H1N1)-like Hemagglutinin (formaldehyde inactivated)|Influenza A virus|Influenza virus type A|Hemagglutinin
- Protein/Peptide Name:Hemagglutinin
- Endotoxin Level:very low
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-110
Specifications
About this item
Recombinant Influenza A virus A/California/7/2009 (H1N1)-like Hemagglutinin Antigen (formaldehyde inactivated).