Influenza A Recombinant Influenza Virus Vaccine
Catalog # 77682-100
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Species:Influenza A
- Protein Synonyms:Influenza virus type A|Influenza Virus Vaccine, Inactivated A-Christchurch-16-2010 NIB-74 (H1N1) (A-California-7-2009) strain|Influenza A virus A/Christchurch/16/2010 NIB-74 (H1N1) Hemagglutinin|Hemagglutinin
- Protein/Peptide Name:Influenza Virus Vaccine
- Endotoxin Level:very low
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-100
Specifications
About this item
Recombinant Influenza A virus A/Christchurch/16/2010 NIB-74 (H1N1) Hemagglutinin Antigen (propiolactone inactivated).