Borrelia Recombinant VisE1 (from E. coli)
Catalog # 77682-092
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Borrelia
- Protein Synonyms:VisE1|variable major protein like sequence E1
- Protein/Peptide Name:VisE1
- Purity:Greater than 95.0% as determined by SDS-page
- Molecular Weight:43 kDa
- Endotoxin Level:very low
- Formulation:Borrelia VisE1 is supplied in 20 mM HEPES buffer pH 8.0, 200 mM NaCl and 20% glycerol.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-092
Specifications
About this item
Recombinant Borrelia VisE1 Antigen, His-tagged.