Borrelia Recombinant p58 immunodominant regions (from E. coli)
77682-084
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Borrelia
- Protein Synonyms:p58 immunodominant regions|p58
- Protein/Peptide Name:p58 immunodominant regions
- Purity:Greater than 80.0% as determined by SDS-page
- Molecular Weight:59815 dalton
- Endotoxin Level:very low
- Formulation:Borrelia p58 is supplied in 20 mM HEPES buffer pH 7.6, 250 mM NaCl and 20% glycerol.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-084
Recombinant Borrelia p58 Antigen, His-tagged.