Borrelia Recombinant p100 immunodominant regions (from Sf9 insect Cells)
77682-076
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:Insect cells (Sf9)
- Species:Borrelia
- Protein Synonyms:p83 immunodominant regions|p83|p100 (p100/p83)|p100|p100 immunodominant regions
- Protein/Peptide Name:p100 immunodominant regions
- Purity:Greater than 80.0% as determined by SDS-page
- Molecular Weight:77813 dalton
- Endotoxin Level:very low
- Formulation:Borrelia p100 (0.77 mg/1 ml) is supplied in 20 mM HEPES buffer pH 8.0, 200 mM NaCl and 20% glycerol.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-076
Recombinant Borrelia p100 Antigen, His-tagged.