Borrelia Garinii Recombinant dbpA (from E. coli)
77682-064
New Product
:
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Borrelia garinii
- Protein Synonyms:dbpA|ATP-dependent RNA helicase DbpA
- Protein/Peptide Name:dbpA
- Purity:Greater than 80.0% as determined by SDS-page
- Molecular Weight:19 kDa
- Endotoxin Level:very low
- Formulation:Borrelia Garinii DbpA is supplied in 20 mM HEPES buffer pH 7.6, 250 mM NaCl and 20% glycerol.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-064
Recombinant Borrelia Garinii DbpA Antigen, His-tagged.