Human Recombinant ACE2 (from HEK293 Cells)
Catalog # 77682-038
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Protein Synonyms:ACEH|angiotensin I converting enzyme (peptidyl-dipeptidase A) 2|angiotensin converting enzyme 2|angiotensin-converting enzyme 2|angiotensin-converting enzyme homolog|ACE2|peptidyl-dipeptidase A|metalloprotease MPROT15|truncated angiotensin converting enzyme 2|angiotensin-converting enzyme-related carboxypeptidase|ACE-related carboxypeptidase|EC 3.4.17.23|angiotensin I converting enzyme 2
- Protein/Peptide Name:ACE2
- Purity:ACE-2 Protein is >95% pure as determined SDS-page
- Endotoxin Level:very low
- Formulation:ACE2 Human protein solution is supplied in 50 mM Tris-HCl, pH 7.5, and 150 mM NaCl and glycerol.
- Grade:Ultrapure grade
- Shipping Temperature:Dry Ice
- Cat. No.:77682-038
Specifications
About this item
Recombinant Human ACE2 Protein, Fc-tagged.