About this item
Activin A is a member of the transforming growth factor beta (TGF-β) family of proteins and functions to stimulate follicle-stimulating hormone (FSH) secretion. Activins are produced in many tissue types including the skin, gonads, lungs, and pituitary gland. Activins interact with receptor type I and type II serine/threonine protein kinases, to activate SMAD signaling and regulate diverse cellular functions, such as cell proliferation, differentiation, wound healing, apoptosis, and metabolism. Activin A is a homodimer comprised of two activin βA chains. Cleavage of the N-terminal propeptide renders the Activin protein biologically active. Human Activin A shares 100% amino acid sequence identity with mouse, rat, porcine, bovine, and feline Activin A proteins.
- High Quality
- Low Endotoxin
- Affordable
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:HumanRatMouse
- Storage Conditions:At or below –20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: Cytotoxicity of MPC-11 cells, ED50 ≤10 ng/ml, Bioactivity ≥1.0×10⁵ units/mg
- Gene ID:P08476 / Q04998 / P18331
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:Inhibin beta-1|FRP|FSH-releasing protein
- Protein/Peptide Name:Activin-A
- Purity:≥95%
- Molecular Weight:13.1/26.2 kDa
- Sequence:MGLECDGKVN ICCKKQFFVS FKDIGWNDWI IAPSGYHANY CEGECPSHIA GTSGSSLSFH STVINHYRMR GHSPFANLKS CCVPTKLRPM SMLYYDDGQN IIKKDIQNMI VEECGCS
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay