Anti-GABRA1 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:25-1:100
Type:
Antigen: GABRA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77706-769
- 77706-770
- Antigen Symbol:
- GABRA1
- GABRA1
- Antigen Synonyms:
- GABA(A) receptor subunit alpha-1|Gamma aminobutyric acid (GABA) A receptor alpha 1|Gamma aminobutyric acid type A receptor alpha1 subunit|GABA(A) receptor|Gamma-aminobutyric acid receptor subunit alpha-1|GABRA1|ECA4|EIEE19|Gamma aminobutyric acid receptor subunit alpha 1|Gaba receptor alpha 1 polypeptide|GBRA1_HUMAN|EJM5|GABA(A) receptor, alpha 1|GABRA 1|Gamma aminobutyric acid A receptor alpha 1|GABA(A) receptor subunit alpha 1|EJM
- GABA(A) receptor subunit alpha-1|Gamma aminobutyric acid (GABA) A receptor alpha 1|Gamma aminobutyric acid type A receptor alpha1 subunit|GABA(A) receptor|Gamma-aminobutyric acid receptor subunit alpha-1|GABRA1|ECA4|EIEE19|Gamma aminobutyric acid receptor subunit alpha 1|Gaba receptor alpha 1 polypeptide|GBRA1_HUMAN|EJM5|GABA(A) receptor, alpha 1|GABRA 1|Gamma aminobutyric acid A receptor alpha 1|GABA(A) receptor subunit alpha 1|EJM
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14867
- P14867
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human GABRA1
- Synthetic peptide of Human GABRA1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together