Anti-UBE2K Rabbit Polyclonal Antibody
New Product
Type:
Antigen: UBE2K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77706-427
- 77706-426
- Antigen Symbol:
- UBE2K
- UBE2K
- Antigen Synonyms:
- ubiquitin conjugating enzyme E2K (UBC1 homolog, yeast)|HYPG|Ubiquitin conjugating enzyme E2 25 kDa|Huntingtin-interacting protein 2|E2 25K|UBE2K_HUMAN|UBC1|Ubiquitin carrier protein|Ubiquitin-conjugating enzyme E2(25K)|Huntingtin interacting protein 2|E2(25K)|UBE2K|LIG|Ubiquitin protein ligase|Ubiquitin-protein ligase|ubiquitin conjugating enzyme E2K|HIP-2|Ubiquitin-conjugating enzyme E2-25K|Ubiquitin-conjugating enzyme E2-25 kDa|Ubiquitin-conjugating enzyme E2 K
- ubiquitin conjugating enzyme E2K (UBC1 homolog, yeast)|HYPG|Ubiquitin conjugating enzyme E2 25 kDa|Huntingtin-interacting protein 2|E2 25K|UBE2K_HUMAN|UBC1|Ubiquitin carrier protein|Ubiquitin-conjugating enzyme E2(25K)|Huntingtin interacting protein 2|E2(25K)|UBE2K|LIG|Ubiquitin protein ligase|Ubiquitin-protein ligase|ubiquitin conjugating enzyme E2K|HIP-2|Ubiquitin-conjugating enzyme E2-25K|Ubiquitin-conjugating enzyme E2-25 kDa|Ubiquitin-conjugating enzyme E2 K
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P61086
- P61086
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ubiquitin-conjugating enzyme E2 K protein (2-200AA)
- Recombinant Human Ubiquitin-conjugating enzyme E2 K protein (2-200AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together