Anti-CTNNA1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200
Type:
Antigen: CTNNA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77706-408
- 77706-407
- Antigen Symbol:
- CTNNA1
- CTNNA1
- Antigen Synonyms:
- NY REN 13 antigen|Ctnna1|CTNNA 1|MDPT2|FLJ52416|CTNA1_HUMAN|OTTHUMP00000224147|Catenin alpha 1|Catenin (cadherin associated protein) alpha 1 102kDa|CAP102|FLJ36832|Renal carcinoma antigen NY REN 13|Catenin alpha-1|OTTHUMP00000224141|Cadherin associated protein 102kDa|Cadherin-associated protein|Cadherin associated protein|CAP 102|Renal carcinoma antigen NY-REN-13|Catenin (cadherin associated protein), alpha 1, 102kDa|Alpha E-catenin
- NY REN 13 antigen|Ctnna1|CTNNA 1|MDPT2|FLJ52416|CTNA1_HUMAN|OTTHUMP00000224147|Catenin alpha 1|Catenin (cadherin associated protein) alpha 1 102kDa|CAP102|FLJ36832|Renal carcinoma antigen NY REN 13|Catenin alpha-1|OTTHUMP00000224141|Cadherin associated protein 102kDa|Cadherin-associated protein|Cadherin associated protein|CAP 102|Renal carcinoma antigen NY-REN-13|Catenin (cadherin associated protein), alpha 1, 102kDa|Alpha E-catenin
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P35221
- P35221
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CTNNA1
- Fusion protein of Human CTNNA1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together