Anti-RETNLB Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RETNLB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77706-273
- 77706-274
- Antigen Symbol:
- RETNLB
- RETNLB
- Antigen Synonyms:
- Retnlb|XCP2|Colon carcinoma-related gene protein|CCGR|RELM beta|Colon carcinoma related gene protein|Cysteine rich secreted protein A12 alpha like 1|Resistin like beta|HXCP2|Colon and small intestine specific cysteine rich protein|C/EBP epsilon regulated myeloid specific secreted cysteine rich protein precursor 2|RETNB_HUMAN|Resistin like protein beta|FIZZ1|Cysteine-rich secreted protein FIZZ2|RELMb|Cysteine rich secreted protein FIZZ2|Cysteine-rich secreted protein A12-alpha-like 1|FIZZ2|RELMbeta|RETNL2|Resistin-like beta|Colon and small intestine-specific cysteine-rich protein|Found in inflammatory zone 1|Cysteine rich secreted A12 alpha like protein 1
- Retnlb|XCP2|Colon carcinoma-related gene protein|CCGR|RELM beta|Colon carcinoma related gene protein|Cysteine rich secreted protein A12 alpha like 1|Resistin like beta|HXCP2|Colon and small intestine specific cysteine rich protein|C/EBP epsilon regulated myeloid specific secreted cysteine rich protein precursor 2|RETNB_HUMAN|Resistin like protein beta|FIZZ1|Cysteine-rich secreted protein FIZZ2|RELMb|Cysteine rich secreted protein FIZZ2|Cysteine-rich secreted protein A12-alpha-like 1|FIZZ2|RELMbeta|RETNL2|Resistin-like beta|Colon and small intestine-specific cysteine-rich protein|Found in inflammatory zone 1|Cysteine rich secreted A12 alpha like protein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BQ08
- Q9BQ08
- Isotype:
- IgG
- IgG
- Immunogen:
- Fusion protein of Human RETNLB
- Fusion protein of Human RETNLB
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together