Anti-Ccl4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: Ccl4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77706-089
- 77689-096
- Antigen Symbol:
- Ccl4
- Ccl4
- Antigen Synonyms:
- Scya4|Macrophage inflammatory protein 1-beta|ACT-2|Mip1b|Small-inducible cytokine A4|MIP-1-beta|Immune activation protein 2|SIS-gamma|Ccl4|C-C motif chemokine 4|ACT2|Protein H400
- Scya4|Macrophage inflammatory protein 1-beta|ACT-2|Mip1b|Small-inducible cytokine A4|MIP-1-beta|Immune activation protein 2|SIS-gamma|Ccl4|C-C motif chemokine 4|ACT2|Protein H400
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14097
- P14097
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Mouse C-C motif chemokine 4 protein (24-92AA)
- Recombinant Mouse C-C motif chemokine 4 protein (24-92AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.1 mol/L NaCl, 15 mmol/L NaN₃, pH 7.2
- 0.1 mol/L NaCl, 15 mmol/L NaN₃, pH 7.2
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together