Anti-SLC27A5 Rabbit Polyclonal Antibody
New Product
Type:
Antigen: SLC27A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77705-953
- 77705-952
- Antigen Symbol:
- SLC27A5
- SLC27A5
- Antigen Synonyms:
- VLACSR|VLACS related|BA-CoA ligase|Very long-chain acyl-CoA synthetase-related protein|Bile acid-CoA ligase|S27A5_HUMAN|Fatty acid Coenzyme A ligase very long chain 3|FATP5|VLCS-H2|BAL|Cholate CoA ligase|Very long chain acyl CoA synthetase homolog 2|FATP-5|FLJ22987|VLCSH2|VLCS H2|Fatty acid transport protein 5|BA CoA ligase|Slc27a5|ACSB|Solute carrier family 27 member 5|FACVL3|Very long chain acyl CoA synthetase related protein|ACSVL6|Very long-chain acyl-CoA synthetase homolog 2|Bile acyl CoA synthetase|very long-chain 3|FATP 5|BACS|Cholate--CoA ligase|VLACS-related|Solute carrier family 27 (fatty acid transporter) member 5|Fatty-acid-coenzyme A ligase|Bile acyl-CoA synthetase|Bile acid CoA ligase
- VLACSR|VLACS related|BA-CoA ligase|Very long-chain acyl-CoA synthetase-related protein|Bile acid-CoA ligase|S27A5_HUMAN|Fatty acid Coenzyme A ligase very long chain 3|FATP5|VLCS-H2|BAL|Cholate CoA ligase|Very long chain acyl CoA synthetase homolog 2|FATP-5|FLJ22987|VLCSH2|VLCS H2|Fatty acid transport protein 5|BA CoA ligase|Slc27a5|ACSB|Solute carrier family 27 member 5|FACVL3|Very long chain acyl CoA synthetase related protein|ACSVL6|Very long-chain acyl-CoA synthetase homolog 2|Bile acyl CoA synthetase|very long-chain 3|FATP 5|BACS|Cholate--CoA ligase|VLACS-related|Solute carrier family 27 (fatty acid transporter) member 5|Fatty-acid-coenzyme A ligase|Bile acyl-CoA synthetase|Bile acid CoA ligase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2P5
- Q9Y2P5
- Isotype:
- IgG
- IgG
- Immunogen:
- Synthetic peptide of Human SLC27A5
- Synthetic peptide of Human SLC27A5
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together