Anti-CYP2C8 Rabbit Polyclonal Antibody
Catalog # 77702-005
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CYP2C8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P10632
- Antigen Synonyms:Microsomal monooxygenase|P450 form 1|Cytochrome P450 IIC2|Cytochrome P450 subfamily IIC (mephenytoin 4 hydroxylase) polypeptide 8|Cytochrome P450 form 1|CYPIIC8|S mephenytoin 4 hydroxylase|P450 MP 12/MP 20|Flavoprotein linked monooxygenase|Cytochrome P450 family 2 subfamily C polypeptide 8|Cytochrome P450 MP-20|CP2C8_HUMAN|Xenobiotic monooxygenase|S-mephenytoin 4-hydroxylase|CPC8|Cytochrome P450 MP-12|Cytochrome P450 2C8|MP 12/MP 20|P450 IIC2|CYP2C8
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human CYP2C8.
- Purification:Antigen affinity purified
- Cat. No.:77702-005
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: CYP2C8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human