Anti-PPP1R1C Rabbit Polyclonal Antibody
Catalog # 77701-977
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:PPP1R1C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q8WVI7
- Antigen Synonyms:IPP5|Inhibitor-5 of protein phosphatase 1|PPP1R1C|Protein phosphatase 1 regulatory subunit 1C
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human PPP1R1C.
- Purification:Antigen affinity purified
- Cat. No.:77701-977
- Packaging:Vial
Specifications
About this item
Type:
Antigen: PPP1R1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse