Anti-EME1 Rabbit Polyclonal Antibody
Catalog # 77701-946
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:EME1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q96AY2
- Antigen Synonyms:MMS4L|essential meiotic endonuclease 1 homolog 1 (S pombe)|Essential meiotic endonuclease 1|Eme1 essential meiotic endonuclease 1 homolog 1 (S. pombe)|MMS4 |6820428D13|MGC106543|EC 3.1.22.-|EME1|MMS4 homolog|essential meiotic endonuclease 1 homolog 2|hMMS4|FLJ31364|S. pombe|1|homolog of yeast EME1 endonuclease|homolog of|EME1_HUMAN|Crossover junction endonuclease EME1|Essential Meiotic Endonuclease 1 Homolog 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Eme1.
- Purification:Antigen affinity purified
- Cat. No.:77701-946
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:5000
Type:
Antigen: EME1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human