Anti-TMEM37 Rabbit Polyclonal Antibody
77701-903
New Product
- Antigen Symbol:TMEM37
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q8WXS4
- Antigen Synonyms:Neuronal voltage-gated calcium channel gamma-like subunit|PRVoltage-dependent calcium channel gamma-like subunit|TMEM37|Transmembrane protein 37
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human TMEM37.
- Purification:Antigen affinity purified
- Cat. No.:77701-903
- Packaging:Vial
IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: TMEM37
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human