Anti-COL6A3 Rabbit Polyclonal Antibody
Catalog # 77701-825
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:COL6A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P12111
- Antigen Synonyms:CO6A3_HUMAN|Collagen type VI alpha 3|Collagen VI alpha 3|COL6A3|Collagen alpha-3(VI) chain
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human COL6A3.
- Purification:Antigen affinity purified
- Cat. No.:77701-825
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:40000
Type:
Antigen: COL6A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse