Anti-SLC24A4 Rabbit Polyclonal Antibody
100 µg, Vial
77701-784
: A19605-100UG
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: SLC24A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Size:100 µg
- Packaging:Vial
- Clonality:Polyclonal
- Purification:Antigen affinity purified
- Form:Liquid
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77701-784
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:Shipped on blue ice
- Antigen Symbol:SLC24A4
- Gene ID:Q8NFF2
- Immunogen:Synthesized peptide derived from the Internal region of Human NCKX4.
- Conjugation:Unconjugated
- Antigen Synonyms:Solute carrier family 24 member 4|SLC24A4|Na(+)/K(+)/Ca(2+)-exchange protein 4|SHEP6|NCKX4|Sodium/potassium/calcium exchanger 4|Solute carrier family 24 (sodium/potassium/calcium exchanger) member 4|NCKX4_HUMAN|SLC24A2
- ELISA:Yes
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



