Anti-MED8 Rabbit Polyclonal Antibody
Catalog # 77701-715
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MED8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q96G25
- Antigen Synonyms:Mediator of RNA polymerase II transcription subunit 8 homolog|MED8_HUMAN|Mediator complex subunit 8|Mediator of RNA polymerase II transcription subunit 8|med8|ARC32|Activator-recruited cofactor 32 kDa component|Activator recruited cofactor 32 kDa component
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Med8.
- Purification:Antigen affinity purified
- Cat. No.:77701-715
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:5000
Type:
Antigen: MED8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat