Anti-TRIM16 Rabbit Polyclonal Antibody
77701-607
New Product
- Antigen Symbol:TRIM16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:O95361
- Antigen Synonyms:Estrogen responsive B box protein|Tripartite motif-containing protein 16|Tripartite motif containing protein 16|Tripartite motif containing 16|TRI16_HUMAN|TRIM16|Estrogen-responsive B box protein|TRIM 16|EBBP
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human TRIM16.
- Purification:Antigen affinity purified
- Cat. No.:77701-607
- Packaging:Vial
IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: TRIM16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human