Anti-USP16 Rabbit Polyclonal Antibody
Catalog # 77701-598
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:USP16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9Y5T5
- Antigen Synonyms:Deubiquitinating enzyme 16|Ubiquitin thiolesterase 16|MSTP039|Ubiquitin carboxyl-terminal hydrolase 16|Ubiquitin Specific Protease 16|usp16|Ubiquitin-specific-processing protease 16|UBP16_HUMAN|Ubiquitin Specific peptidase 16|UBP M|Human ubiquitin processing protease|UBPM|Ubiquitin-processing protease UBP-M
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human USP16.
- Purification:Antigen affinity purified
- Cat. No.:77701-598
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: USP16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat