Anti-CLTCL1 Rabbit Polyclonal Antibody
Catalog # 77687-419
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CLTCL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P53675
- Antigen Synonyms:CLH-22|CLTCL|CLTCL1|CLTDClathrin heavy chain 2|Clathrin heavy chain on chromosome 22|CLH22
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Clathrin heavy chain 2 protein (1423-1566AA)
- Purification:>95%, Protein G purified
- Cat. No.:77687-419
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: CLTCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse